Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Nitroprusside disodium dihydrate | 13755-38-9 | 99.6% | C5H4FeN6Na2O3 | 1 ML
SDP

Supplier:  Medchemexpress LLC HYA011910MM/1ML

Encompass_Preferred

Nitroprusside disodium dihydrate is a vasodilator used for research into acute hypertension and heart failure. It induces autophagy in glutathione-depleted osteoblasts and acts as a nitric oxide (NO) donor in a rat intestinal ischemia reperfusion model. It also sensitizes human gastric cancer cells to tumor necrosis factor-related apoptosis-inducing ligand (TRAIL)-induced apoptosis.

  • Solid appearance
  • Reddish brown to dark brown color
  • Ships at room temperature
  • Store at 4°C, sealed, away from moisture and light
  • Soluble in water and DMSO
  • For research use only

Catalog No. 50-004-32348


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.