Learn More
Medchemexpress LLC Nitroprusside disodium dihydrate | 13755-38-9 | 99.62% | C5H4FeN6Na2O3 | 500 MG
SDP
Supplier:  Medchemexpress LLC HYA0119500MG
Encompass_Preferred
Nitroprusside disodium dihydrate is a vasodilator used in research for acute hypertension and heart failure. This compound induces autophagy, acts as a nitric oxide (NO) donor, and has shown activity in sensitizing human gastric cancer cells to apoptosis.
  • Vasodilator for acute hypertension research
  • Used in heart failure research
  • Induces autophagy in glutathione-depleted osteoblasts
  • Acts as a nitric oxide (NO) donor
  • Sensitizes human gastric cancer cells to TRAIL-induced apoptosis
Catalog No. 50-004-33567