Learn More
ATCC Nitrosomonas europaea
Supplier:  ATCC 19718
Encompass
Genome sequencing strain. Depositor states that this strain produces hydroxylamine oxidase [hydroxylamine oxidoreductase].
Catalog No. 50-238-4071