Learn More
GA International NitroTAG – Cryogenic Barcode Labels for Liquid Nitrogen Storage
Supplier:  GA International JTTA118C34WH
Encompass_Preferred
These labels are ideally suited for storage of small diameter cryo tubes and vials, polypropylene cryo/freezer boxes, cardboard freezer boxes, cell culture plates, and plastic bags in liquid and vapor phase liquid nitrogen (-196°C/-321°F), and lab freezers (-80°C/-112°F), as well as for dry ice transportation. Waterproof and made of a strong thermoplastic film, these labels can be immersed in liquid nitrogen immediately following application, and withstand multiple freeze-thaw cycles. Compatible with all common thermal-transfer printers, they can be printed on with alphanumeric text, barcodes, serial numbers, serial barcodes, and other types of information.
Catalog No. NC1630685