Learn More
ALADDIN SCIENTIFIC CORPORATION Nitrotetrazolium Blue chloride, 1g
Supplier:  ALADDIN SCIENTIFIC CORPORATION N1049081G
Encompass_Preferred
p-Nitroblue Tetrazolium Chloride is an NADPH-diaphorase substrate that competitively inhibits NOS (nitric oxide synthase) (IC50 3-4 uM) p-Nitroblue Tetrazolium Chloride is a well-known scavenger of superoxide anions and is useful as a substrate for alkaline phosphatase often in conjunction with BCIP Substrate for dehydrogenases and other oxidases
Catalog No. 50-000-82553