Learn More
Perkin Elmer US LLC Nitrous Oxide Regulator, Heated, 115 V
Supplier:  Perkin Elmer US LLC 03030204
Encompass_Preferred
The regulator comes with a nitrous oxide line (00470258) and tubing adapters for 1/4 and 5/16 tubing on outlet side. The 220 V Nitrous Oxide Regulator (03030349) is available separately.
Catalog No. 50-211-2752