Learn More
Medchemexpress LLC Nitroxynil | 1689-89-0 | MFCD00070776 | 98.7% | 290.016 g/mol | C7H3IN2O3 | 25 G
SDP
Supplier:  Medchemexpress LLC 25GHYW049875
Encompass_Preferred
Nitroxynil is an anthelmintic compound used as an analytical standard in research on trematode infections such as Fasciola hepatica. It is supplied as a solid analytical standard intended for laboratory and research applications.
  • Used in research of Fasciola hepatica and other trematode infections.
  • High purity (98.7%).
  • Chemical formula C7H3IN2O3 and molecular weight 290.016 g/mol.
  • Packaged as a 25 G solid for analytical use.
Catalog No. 50-002-08560