Learn More
Bioworld Nitsch Vitamin Concentrate (Syringe Vials), 100 mL
Supplier:  Bioworld 306300971
Encompass_Preferred
Nitsch Vitamin ConcentrateContains Vitamins as described by Nitsch & Nitsch (1969)Package includes 10 prepared, filtered, and sterilized ampules of 10 mL of 100X vitamin stock solution Use 1 mL per liter of mediumContents: Biotin: 0.05 mg/L Folic acid: 0.50 mg/L Glycine: 2.00 mg/L myo Inositol: 100.0 mg/L Nicotinic acid: 5.0 mg/L Pyridoxine HCl: 0.5 mg/L Thiamine HCl: 0.5 mg/L
Catalog No. 50-254-756