Learn More
Sino Biological Nipah virus Fusion glycoprotein (His Tag)
Supplier:  Sino Biological 40982V08H100
Encompass_Preferred
A DNA sequence encoding the NIV Fusion glycoprotein (QKV44013.1) (Ile27-Asp482) was expressed, with a polyhistidine tag at the C-terminus.
Catalog No. 50-299-3604