Learn More
Sino Biological Nipah virus Glycoprotein G Protein (ECD, His Tag)
Supplier:  Sino Biological 40980V08H100
Encompass_Preferred
A DNA sequence encoding the NIV Glycoprotein G (AAY43916.1) (Gln71-Thr602) was expressed, with a polyhistidine tag at the C-terminus.
Catalog No. 50-255-3568