Learn More
Medchemexpress LLC NKG2DL2 Human HEK2 50ug

Supplier: Medchemexpress LLC HYP7116450UG
Recombinant human NKG2DL2 extracellular domain (residues G26-S217), expressed in HEK293 cells and supplied lyophilized with a C-terminal 6xHis tag. The protein activates the KLRK1/NKG2D receptor, does not bind β2-microglobulin, and is provided at high purity for use in NK cell biology, receptor-ligand interaction studies, and related research applications.
- Expressed in HEK293 cells with a C-terminal His tag.
- Contains residues G26-S217 representing the extracellular domain.
- High purity (>95%) as determined by reducing SDS-PAGE.
- Activates KLRK1/NKG2D receptor for NK cell functional assays.
- Lyophilized formulation for stable, long-term storage.
- Reconstitution guidance: do not reconstitute below 100 μg/mL; aliquot and freeze for extended storage.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.