Learn More
Medchemexpress LLC HY-103193 10mg , NKH477 CAS:138605-00-2 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY10319310MG
Encompass_Preferred
HY-103193 10mg NKH477 CAS: 138605-00-2 NKH477 (Colforsin dapropate hydrochloride) directly activates the catalytic unit of adenylate cyclase and increases intracellular cAMP. NKH477 is a forskolin derivative that improves cardiac failure mainly through its beneficial effects on diastolic cardiac function. NKH477 exerts an antiproliferative effect in vivo with an altered cytokine profile to inhibit the acute rejection of rat orthotopic lung allografts. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-226-0439