Learn More
Genscript Corporation NKp30/NCR3/CD337Biotin His
Supplier:  Genscript Corporation Z04570100
Encompass
NKp30 along with NKp44 and NKp46 constitute a group of receptors termed Natural Cytotoxicity Receptors These receptors play a major role in triggering NK-mediated killing of most tumor cells lines NKp30 stimulates NK cells cytotoxicity toward neighboring cells producing these ligands It controls for instance NK cells cytotoxicity against tumor cells
Catalog No. 50-000-57124