Learn More
Medchemexpress LLC NKp30/NCR3 Rat HEK 50ug | 50ug
SDP
Supplier:  Medchemexpress LLC HYP7710650UG
Encompass_Preferred
NKp30/NCR3 protein is a cell membrane receptor on natural killer (NK) cells that activates cytotoxicity by binding to ligands such as BAG6 and NCR3LG1 stimulating NK cells to respond against neighboring cells (including tumor cells) expressing these ligands NKp30/NCR3 Protein Rat (HEK293 His) is the recombinant rat-derived NKp30/NCR3 protein expressed by HEK293 with C-His labeled tag
Catalog No. 50-001-85593