Learn More
Medchemexpress LLC NKp80/KLRF1 Human 100ug
SDP
Supplier:  Medchemexpress LLC HYP72501100UG
Encompass_Preferred
The NKp80/KLRF1 protein is critically involved in NK-mediated PHA-induced cytolysis of lymphoblasts emphasizing its importance in immune responses against activated cells Its involvement suggests a role in identifying and targeting specific cellular targets for elimination NKp80/KLRF1 Protein Human (HEK293 His) is the recombinant human-derived NKp80/KLRF1 protein expressed by HEK293 with N-6 His labeled tag
Catalog No. 50-001-81377