Learn More
Cayman Chemical NKRP1ACD161 EXrAClular D 100ug
SDP
Supplier:  Cayman Chemical 38063100 UG
Encompass_Preferred
Source: Recombinant mouse N-terminal His-tagged NKR-P1A extracellular domain expressed in HEK293 cells • Amino acids: 67-277 • MW: 21 kDa
Catalog No. 50-291-7462