Learn More
Medchemexpress LLC HY-135231 10mg Medchemexpress, NL-1 CAS:188532-26-5 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY13523110MG
Encompass_Preferred
Medchemexpress, HY-135231 10mg NL-1 CAS:188532-26-5 NL-1 is a mitoNEET inhibitor with antileukemic effect. NL-1 inhibits REH and REH/Ara-C cells growth with IC50s of 47.35 µM and 56.26 µM, respectively. NL-1-mediated death in leukemic cells requires the activation of the autophagic pathway. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-203-0328