Learn More
Selleck Chemical LLC NL-1 S0767-10MG
SDP
Supplier:  Selleck Chemical LLC S076710MG
Encompass_Preferred
The mitochondrial mitoNEET ligand NL-1 is a mitoNEET inhibitor which induces a concentration-dependent decrease in cell viability with an IC50 of 47 35 M and 56 26 M in REH and REH/Ara-C cells respectively The docking studies using liver mitochondrial suspensions show the IC50 of NL-1 binding for [3H]-rosiglitazone is 0 9 M and the Ki for NL-1 site 1 and site 2 was 4 78 and 2 77 respectively
Catalog No. 50-000-42579