Learn More
Cayman Chemical nlfInavIrmesylate 5mg
SDP
Supplier:  Cayman Chemical 151445 MG
Encompass_Preferred
An orally bioavailable inhibitor of the HIV-1 protease (Ki =2 nM) that is used in combination with HIV reverse transcriptase inhibitors for the treatment of HIV infection
Catalog No. 50-296-9162