Learn More
Selleck Chemical LLC IDO-IN-2 10mg 1402836-58-1 RG 6078, NLG-919 analogue, GDC-0919 analogue
SDP
Supplier:  Selleck Chemical LLC S711110MG
Encompass_Preferred
IDO-IN-2 (RG 6078, NLG-919 analogue, GDC-0919 analogue) is a a potent IDO1 inhibitor with IC50 of 38 nM. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-194-7972