Learn More
Apexbio Technology LLC NLG919, 10mM (in 1mL DMSO). Cas: 1402836-58-1 MFCD: MFCD26142661. Potent IDO pathway inhibitor
SDP
Supplier:  Apexbio Technology LLC A43735.1
Encompass_Preferred
NLG919 is a novel and orally-bioavailable small-molecule inhibitor of indoleamine 2,3-dioxygenase (IDO) pathway, a crucial pathway involved in allergic inflammation that mediates immunosuppressive effects through metabolization of tryptophan (Trp) to kynurenine. Other sizes are available. Please inqury us for quote.
Catalog No. 50-190-4933