Learn More
ECM Bioscience NLRC4 (Ser-533), phospho-specific Rabbit Polyclonal Antibody, ECM Biosciences
Supplier:  ECM Bioscience NP5411
Encompass_Preferred
The antibody was cross-adsorbed to unphosphorylated NLRC4 (Ser-533) peptide before affinity purification using phospho-NLRC4 (Ser-533) peptide. This antibody antibody detects a 110 kDa* protein on SDS-PAGE immunoblots of PMA-differentiated THP1 human macrophages. In addition, the antibody detects a mouse recombinant phospho-NLRC4 (Ser-533) peptide, but does not detect this peptide after lambda phosphatase treatment. The immunogen was Phospho-NLRC4 (Ser-533) synthetic peptide (coupled to carrier) corresponding to amino acids surrounding serine 533 in mouse NLRC4. This sequence has high homology to human and rat NLRC4, and is not homologous to sequences in other NLR family members. Accession #Q3UP24. Applications = WB, E; Species Reactivity = Hu, Rt, Ms; Size = 100 μl; Buffer = PBS, 50% glycerol, 1 mg/ml BSA, and 0.05% sodium azide. Store at –20°C. Stable for 1 year. Other Names = CARD12, CLAN1, IPAF, NLR family CARD domain-containing protein 4, NOD-like receptor 4
Catalog No. 50-172-0835