Learn More
ABclonal Technology NLRP12 Rabbit pAb
SDP
Supplier:  ABclonal Technology A14519500UL
Encompass_Preferred
This gene encodes a member of the CATERPILLER family of cytoplasmic proteins. The encoded protein, which contains an N-terminal pyrin domain, a NACHT domain, a NACHT-associated domain, and a C-terminus leucine-rich repeat region, functions as an attenuating factor of inflammation by suppressing inflammatory responses in activated monocytes. Mutations in this gene cause familial cold autoinflammatory syndrome type 2. Alternative splicing results in multiple transcript variants.
Catalog No. 50-001-09069