Learn More
Medchemexpress LLC Nls (pkkkrkv) | 95088-49-6 | 99.0% | 883.14 | 10 MG
SDP
Supplier:  Medchemexpress LLC HYP187610MG
Encompass_Preferred
NLS (PKKKRKV) is a nuclear localization signal derived from the SV40 large T antigen. It mediates the binding of karyophilic proteins to importin α and is used to enhance nuclear entry in gene transfer research.
  • Derived from simian virus 40 large T antigen
  • Mediates binding of karyophilic protein to importin α
  • Enhances nuclear entry in gene transfer research
  • High purity (99.0%)
  • Appears as a white to off-white solid
Catalog No. 50-004-24377