Learn More
Medchemexpress LLC NLS (PKKKRKV) hydrochloride | 99.02% | 919.60 | 5 MG
SDP
Supplier:  Medchemexpress LLC 5MGHYP1876B
Encompass_Preferred
NLS (PKKKRKV) hydrochloride is the hydrochloride form of NLS (PKKKRKV). It is a nuclear localization signal (NLS) derived from the simian virus 40 large tumor antigen (SV40 large T antigen), which mediates the binding of karyophilic protein to importin α. It can be used to enhance nuclear entry in gene transfer research.
  • Derived from SV40 large T antigen
  • Mediates binding of karyophilic protein to importin α
  • Enhances nuclear entry in gene transfer research
Catalog No. 50-003-93366