Learn More
Electron Microscopy Sciences NMA (Nadic Methyl Anhydride) 450 ML

Supplier: Electron Microscopy Sciences 19001
(Methyl-5-Norbornene-2,3-Dicarboxylic Anhydride)
CAS #25134-21-8 C₁₀H₁₀O₃ F.W. 178.19
Boiling point: 140°C
An epoxy hardener.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.