Learn More
Cayman Chemical nmadIpIn A 50mg
SDP
Supplier:  Cayman Chemical 2168950 MG
Encompass_Preferred
A phenolic acid and fungal metabolite predicted to inhibit AB17-42 dimerization disrupt preformed AB17-42 trimers and bind to the catalytic site of PDE5A based on molecular dynamics simulation studies
Catalog No. 50-296-4037