Learn More
Lonza Walkersville NHBE – Human Bronchial/Tracheal Epithelial Cells with Retinoic Acid
Supplier:  Lonza Walkersville CC2540
Encompass
Normal and Diseased Bronchial Epithelial (NHBE and DHBE) cells are isolated from epithelial lining of airways above bifurcation of the lungs. Lonza's Normal Human Bronchial Epithelial Cells (NHBE) are available with retinoic acid. All Cryopreserved human bronchial epithelial cells are guaranteed through 15 population doublings. Cells test negative for mycoplasma, bacteria, yeast, and fungi. HIV-1, hepatitis B and hepatitis C are not detected for all donors and/or cell lots. A Certificate of Analysis is provided for each cell lot purchased.Benefits:All cells test negative for mycoplasma, bacteria, yeast, and fungiHIV-1, Hepatitis B and Hepatitis C are not detected for all donors and/or cell lotsCertificate of Analysis provided for every cell lot Content:1 x Cryopreserved ampule NHBE w/ RA containing ≥500,000 cells
Catalog No. NC9192018