Learn More
ABclonal Technology NMNAT2 Rabbit pAb
SDP
Supplier:  ABclonal Technology A1383320UL
Encompass_Preferred
This gene product belongs to the nicotinamide mononucleotide adenylyltransferase (NMNAT) enzyme family, members of which catalyze an essential step in NAD (NADP) biosynthetic pathway. Unlike the other human family member, which is localized to the nucleus, and is ubiquitously expressed, this enzyme is cytoplasmic, and is predominantly expressed in the brain. Two transcript variants encoding different isoforms have been found for this gene.
Catalog No. 50-000-98231