Learn More
Cole-Parmer Wilmad-LabGlass WG-1000-7 NMR Tube, disposable, 7"L, 100 MHz, 100/pk
Supplier:  Cole-Parmer 8399958
Encompass
NMR tubes are precision machined by melting, grinding, and polishing to meet exact tolerances. Each 5mm tube meets precise wall thickness, exact centerline symmetry, and perfect straightness.
Catalog No. NC0480565