Learn More
TARGETMOL CHEMICALS INC NMS-873 10MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T185310MG
Encompass_Preferred
Also available in 1 mL, 2 mg, 5 mg, 25 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. NMS-873 is a potent and selective allosteric inhibitor of VCP/p97. Purity 99.05%
Catalog No. 50-287-8399