Learn More
Medchemexpress LLC NNC-711 hydrochloride | 145645-62-1 | MFCD00153853 | 99.8% | 386.87 g/mol | C21H23ClN2O3 | 50 MG
SDP
Supplier:  Medchemexpress LLC HY10350650MG
Encompass_Preferred
NNC-711 hydrochloride is a potent, selective inhibitor of the GABA transporter GAT-1 used as a research reagent in neuroscience. It exhibits anticonvulsant and analgesic effects in vivo and is commonly used to study GABA uptake and synaptic inhibition.
  • Potent, selective inhibitor of GAT-1 with an IC50 of 0.04 μM (hGAT-1).
  • Hydrochloride salt provided as a solid for laboratory use.
  • Molecular weight 386.87 g/mol and formula C21H23ClN2O3.
  • High purity (~99.8% as listed by the supplier).
  • Supplied as a 50 MG research quantity suitable for in vitro and in vivo studies.
Catalog No. 50-002-46129