Learn More
Abcam NNGH, Matri x metalloprotease (MMP) inhibitor, 25MG
Supplier:  Abcam AB14125925MG
Encompass_Preferred
MW 316.38 Da, Purity >98%. MMP3 (Stromelysin-1) inhibitor. Additionally shows broad spectrum inhibition activity at other MMPs. Cell permeable.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-6755