Learn More
American Radiolabeled Chemicals Inc NNK 3H 50UC
SDP
Supplier:  American Radiolabeled Chemicals Inc ART 177450UC
Encompass
(R,S)-NNK-pyridyl-5-3H 50uCi in ethanol:water (1:1), 1 mCi/ml, S.A. 5-15 Ci/mmol, M.W. 207.2, Shipped in Dry Ice
Catalog No. 50-901-003
Add to Cart
Request a Quote