Learn More
TRIBIOSCIENCE INC Nicotinamide N-methyltransferase (NNMT) Activity Fluorometric Assay Kit, 100 Tests
SDP
Supplier:  TRIBIOSCIENCE INC TBS2098100
Encompass_Preferred
NNMT Activity Fluorometric Assay provides the easiest and most accurate approach to measure NNMT activity in a variety of samples. The free thiol group of the homocysteine can be detected with a thiol probe generating an enhanced fluorescent signal at Ex/Em = 400/465 nm.
Catalog No. NC2886159