Learn More
TRIBIOSCIENCE INC Nicotinamide N-methyltransferase (NNMT) Inhibitor Screening Assay Kit, 100 Tests
SDP
Supplier:  TRIBIOSCIENCE INC TBS2097100
Encompass_Preferred
NNMT inhibitor screening kit provides the easiest, most accurate, and simplest approach for high-throughput screening of NNMT inhibitors. The free thiol group of the homocysteine can be detected with a thiol probe generating an enhanced fluorescent signal at Ex/Em = 400/465 nm.
Catalog No. NC2887476