Learn More
Medchemexpress LLC HY-131042 5mg Medchemexpress, NNMTi CAS:42464-96-0 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1310425MG
Encompass_Preferred
Medchemexpress, HY-131042 5mg NNMTi CAS:42464-96-0 NNMTi is a potent nicotinamide N-methyltransferase (NNMT) inhibitor (IC50=1.2 μM) and selectively binds to the NNMT substrate-binding site residues. NNMTi  promotes myoblast differentiation in vitro and enhances fusion and regenerative capacity of muscle stem cells (muSCs) in aged mice. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-1259