Learn More
Cayman Chemical NnonoylLHomoserIn lacto 10mg
SDP
Supplier:  Cayman Chemical 1686810 MG
Encompass_Preferred
A bacterial quorum-sensing signaling molecule; induces GFP production in E. coli expressing the V. fischeri LuxR-PluxI quorum-sensing operon from 100-1,000 nM
Catalog No. 50-296-7710