Learn More
Gilson Company Inc NO. 10 TEST SCREEN TRAY
Supplier:  Gilson Company Inc TSA101 #10
Encompass_Preferred
No. 10 (2.0mm) full ASTM Testing Screen Trays feature welded and bolted heavy gauge painted steel construction and are fitted with ASTM E11 Compliance Grade wire cloth.
Catalog No. NC2032703