Learn More
Ace Glass, Inc. 25mm Ace-Thred nylon bushing, no O-ring, accepts 24-25mm OD tubing
SDP
Supplier:  Ace Glass, Inc. 750609
Encompass_Preferred
25mm Ace-Thred nylon bushing, no O-ring, accepts 24-25mm OD tubing
Catalog No. 50-974-962