Learn More
Ace Glass, Inc. #25 Ace-Thred solid nylon plug, 7855-734 FETFE O-ring front seal
SDP
Supplier:  Ace Glass, Inc. 584616
Encompass_Preferred
#25 Ace-Thred solid nylon plug with front seal O-ring, for sealing column ends. Permits preparation and storage of columns. Supplied with size -212 FETFE O-ring.
Catalog No. 50-972-333