Learn More
Ace Glass, Inc. 50mm ID x 30cm chromatography column, #3 stopper top, 500mLreservoir, without filter disc
SDP
Supplier:  Ace Glass, Inc. 590724
Encompass_Preferred
50mm I.D. x 30cm chromatography column with 500mL reservoir at top and 2mm bore 1:5 PTFE stopcock plug at bottom. Opening at top is for a #3 stopper.
Catalog No. 50-972-590