Learn More
Accuflow Systems Saint-Gobain Silicone Stopper, Size 3, Platinum-Cured Silicone, Autoclavable
Supplier:  Accuflow Systems 41201003
Encompass_Preferred
Sani-Tech silicone stopper size 3 is an ultra-pure, platinum-cured silicone closure engineered for laboratory and bioprocess applications. It offers high purity, resistance to cracking and spallation, and is fully autoclavable for sterile processing.
  • Ultra-pure platinum-cured silicone.
  • Size 3, suitable for carboys and solution bottles.
  • Dimensions: A 0.950 in (24.1 mm), B 0.725 in (18.4 mm), H 0.982 in (24.9 mm).
  • Fully autoclavable and sterilizable.
  • Resists cracking and spallation, minimizes particulate release.
  • Common applications include process and storage procedures and laboratory apparatus.
Catalog No. NC3314139