Learn More
Ace Glass, Inc. #7-#11 feed tube with hose connection
SDP
Supplier:  Ace Glass, Inc. 582720
Encompass_Preferred
Feed tube adapter with #7 to #11 Ace-Thred ends and hose connection on side arm. #7 thread at top is supplied with a nylon bushing and FETFE o-ring, for use with adjustable feed tubes.
Catalog No. 50-972-217