Learn More
Ace Glass, Inc. Column, 12" ID x 48", 6" O-ring grooved flat flange top, #80 Ace-Thred bottom
SDP
Supplier:  Ace Glass, Inc. 586230
Encompass_Preferred
12 I.D. x 48 chromatography column with a size 6 o-ring grooved flat flange top and #80 Ace-Thred bottom. Supplied with silicone o-ring.
Catalog No. 50-972-393