Learn More
Ace Glass, Inc. Column, 6" ID x 24" , 6" O-ring grooved flat flange top, #80 Ace-Thred bottom
SDP
Supplier:  Ace Glass, Inc. 586212
Encompass_Preferred
6 I.D. x 24 chromatography column with a size 6 o-ring grooved flat flange top and #80 Ace-Thred bottom. Supplied with silicone o-ring.
Catalog No. 50-972-388