Learn More
Medchemexpress LLC No-losartan A | 791122-48-0 | 98.3% | 10 MG

Supplier: Medchemexpress LLC HY12097910MG
No-losartan A is a losartan-derived nitric oxide (NO) donor research compound that releases nitric oxide to produce vasorelaxation and antagonizes angiotensin II-induced vasoconstriction via activity at the angiotensin receptor.
- Displays vasorelaxing effects through nitric oxide release
- Acts as an angiotensin receptor antagonist
- Molecular formula C30H28ClN7O5, molecular weight 602.04 g/mol
- Reported purity approximately 98.3% for research use
- Provided as a small research pack suitable for in vitro and in vivo probe studies
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.