Learn More
HARVARD BIOSCIENCE INC NO1 GLASS COVERSLIP 100/PK
Supplier:  HARVARD BIOSCIENCE INC 640703
Encompass_Preferred
Total order minimum requirement of $500. If only item on order, must order QTY of 11.Coverslip, Thickness No.1: 0.15mm, 15mm D, Compatible Chamber(s): RC-20, RC-20H, RC-25F, for microscopy and imaging
Catalog No. NC9060588