Learn More
Chemglass Life Sciences Cap, GL-14, With Aperture, PBTE
SDP
Supplier:  Chemglass Life Sciences CG19501
Encompass_Preferred
GL-14 Cap with 9.5mm Aperture. Open top cap for use with GL screw threads. The cap combined with the CG-197 sealing ring will accommodate tubing of the listed diameter. Cap is made from PBT (thermoplastic polyester) and will withstand temperatures from -45 to 200¿ C.
Catalog No. CG19501