Learn More
Roboz Surgical NO3 DUMONT DUMOSTAR
SDP
Supplier:  Roboz Surgical RS4966
Encompass
Dumont Tweezers; Pattern #3; .08 X .04mm Tip Size; 120mm Length; Material: Dumostar; Biologie Tips
Catalog No. NC9208914